Learn More
Medchemexpress LLC N-acetyl-L-aspartic acid-d3 | 284665-15-2 | 99.8% | 178.16 | C6H6D3NO5 | 5 MG
SDP
Catalog No. 5000263577
Encompass_Preferred
Deuterium-labeled N-acetyl-L-aspartic acid (NAA-d3) is a stable isotope analytical standard used as a tracer or internal standard for quantitative analysis in NMR, GC-MS, and LC-MS. It is intended for research into brain metabolism and related neurodegenerative disorders and is supplied as a high-purity solid for analytical workflows.
  • Deuterium-labeled internal standard for quantitative analysis
  • High chemical and isotopic purity suitable for sensitive assays
  • White to off-white solid for convenient handling
  • Available in small milligram pack sizes for analytical use
  • Suitable as a tracer in studies of brain metabolism and disease
Catalog No. 50-002-63577 Supplier Medchemexpress LLC Supplier No. HY113524S5MG
Deuterium-labeled N-acetyl-L-aspartic acid (NAA-d3) is a stable isotope analytical standard used as a tracer or internal standard for quantitative analysis in NMR, GC-MS, and LC-MS. It is intended for research into brain metabolism and related neurodegenerative disorders and is supplied as a high-purity solid for analytical workflows.
  • Deuterium-labeled internal standard for quantitative analysis
  • High chemical and isotopic purity suitable for sensitive assays
  • White to off-white solid for convenient handling
  • Available in small milligram pack sizes for analytical use
  • Suitable as a tracer in studies of brain metabolism and disease