Learn More
Alkali Scientific n Acetyl L cysteine, 1 Kg
SDP
Catalog No. 5000660349
Encompass_Preferred
N-acetyl-L-cysteine (NAC) is a widely used antioxidant that serves as a precursor to glutathione, one of the body's most important natural antioxidants. NAC plays a crucial role in protecting cells from oxidative stress and is used in studies related to cellular protection, aging, and disease prevention. It is particularly valuable in the research of liver health, acetaminophen poisoning, and inflammatory conditions. The 1kg quantity of NAC is ideal for high-throughput research where large volumes are required. This form of NAC is also used in laboratory experiments to prevent oxidative damage during cell culture, protein studies, and in vivo models of oxidative stress-related diseases.
Catalog No. 50-006-60349 Supplier Alkali Scientific Supplier No. CET005CP
N-acetyl-L-cysteine (NAC) is a widely used antioxidant that serves as a precursor to glutathione, one of the body's most important natural antioxidants. NAC plays a crucial role in protecting cells from oxidative stress and is used in studies related to cellular protection, aging, and disease prevention. It is particularly valuable in the research of liver health, acetaminophen poisoning, and inflammatory conditions. The 1kg quantity of NAC is ideal for high-throughput research where large volumes are required. This form of NAC is also used in laboratory experiments to prevent oxidative damage during cell culture, protein studies, and in vivo models of oxidative stress-related diseases.