Learn More
Research Products International Corp n-Acetyl-L-cysteine, 1 KG
SDP
Supplier:  Research Products International Corp A100401000.0
Encompass_Preferred
Increases cellular pools of free radical scavenger. Prevents apoptotic death of neuronal cells. Induces apoptosis in human and rat smooth muscle cells.
Pyrogen free.
Catalog No. 50-223-0247