Learn More
Research Products International Corp n-Acetyl-L-cysteine, 100 Grams
SDP
Supplier:  Research Products International Corp A10040100.0
Encompass_Preferred
Increases cellular pools of free radical scavenger. Prevents apoptotic death of neuronal cells. Induces apoptosis in human and rat smooth muscle cells.
Pyrogen free.
Catalog No. 50-213-096