Learn More
Sigma Aldrich Fine Chemicals Biosciences N-Acetyl-L-cysteine, 616-91-1, MFCD00004880, 5G
Catalog No. NC1618628
Encompass
Molecular Formula: HSCH2CH(NHCOCH3)CO2H, Molecular Weight: 163.19, Assay:≥99% (TLC), Solubility: H2O: 100 mg/mL (with heating)
Catalog No. NC1618628 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A91655G
Molecular Formula: HSCH2CH(NHCOCH3)CO2H, Molecular Weight: 163.19, Assay:≥99% (TLC), Solubility: H2O: 100 mg/mL (with heating)