Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-cysteine (Crystalline), MP Biomedicals

Catalog No. ICN19460380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19460380 100 g
ICN19460305 5 g
ICN19460325 25 g
ICN19460350 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19460380 Supplier MP Biomedicals, Inc Supplier No. 0219460380
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Color White
Alpha Vector Acetyl-L-cysteine
Quantity 100 g
Formula Weight 163.2
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Assay ≥96%
Chemical Name or Material N-Acetyl-L-cysteine
EINECSNumber 210-498-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.