Learn More
Medchemexpress LLC HY-134495 100mg , N-Acetyl-L-cysteine ethyl ester CAS:59587-09-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134495100MG
Encompass_Preferred
HY-134495 100mg N-Acetyl-L-cysteine ethyl ester CAS: 59587-09-6 N-Acetyl-L-cysteine ethyl ester is an esterified form of N-acetyl-L-cysteine (NAC). N-Acetyl-L-cysteine ethyl ester exhibits enhanced cell permeability, and produce NAC and cysteine. N-Acetyl-L-cysteine ethyl ester increases circulating hydrogen sulfide (H2S) and can be used as an H2S producer. N-Acetyl-L-cysteine ethyl ester has the potential to substitute NAC as a mucolytic agent, and as a GSH-related antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1386