Learn More
TARGETMOL CHEMICALS INC N-Acetyl-L-cysteine ethyl este
SDP
Catalog No. 502876413
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-Acetyl-L-cysteine ethyl ester (NACET) is a derivative of the amino acid L-cysteine. NACET has been shown to exhibit diverse biochemical and physiological effects, including inhibition of protein synthesis, inhibition of DNA replication, and induction of apoptosis. Additionally, it has demonstrated anti-inflammatory, antioxidant, and anti-cancer activity.
Catalog No. 50-287-6413 Supplier TARGETMOL CHEMICALS INC Supplier No. T4128550MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-Acetyl-L-cysteine ethyl ester (NACET) is a derivative of the amino acid L-cysteine. NACET has been shown to exhibit diverse biochemical and physiological effects, including inhibition of protein synthesis, inhibition of DNA replication, and induction of apoptosis. Additionally, it has demonstrated anti-inflammatory, antioxidant, and anti-cancer activity.