Learn More
Cayman Chemical N-acetyl-L-Glutamic Acid, Purity: ≥90%, CAS: 1188-37-0, 100 g
SDP
Catalog No. 502917920
Encompass_Preferred
An endogenous metabolite; an allosteric activator of mitochondrial carbamoyl phosphate synthetase; the first intermediate in arginine biosynthesis in bacteria; inhibits human liver NAGS (Ki = 0.25 mM); inhibits IDH2, mitochondrial complex IV, and GDH, in rat brain mitochondrial preparations at 1 mM; increased urine levels are associated with ACY1 mutations
Catalog No. 50-291-7920 Supplier Cayman Chemical Supplier No. 37359100 G
An endogenous metabolite; an allosteric activator of mitochondrial carbamoyl phosphate synthetase; the first intermediate in arginine biosynthesis in bacteria; inhibits human liver NAGS (Ki = 0.25 mM); inhibits IDH2, mitochondrial complex IV, and GDH, in rat brain mitochondrial preparations at 1 mM; increased urine levels are associated with ACY1 mutations