Learn More
Medchemexpress LLC N-acetyl-L-glutamic acid | 1188-37-0 | MFCD00002802 | 98.0% | 189.17 g/mol | C7H11NO5 | 1 ML
SDP
Catalog No. 5000303208
Encompass_Preferred
N-Acetyl-L-glutamic acid is an endogenous N-acyl derivative of L-glutamic acid used as a biochemical reagent and analytical standard. The product is available as a ready-to-use 10 mM solution in DMSO (1 mL) and is accompanied by molecular data and solubility guidance for laboratory use.
  • Endogenous metabolite involved in the urea cycle and metabolic assays.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and screening.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular weight 189.17 g/mol and formula C7H11NO5 for reference.
  • Recommended solvent and formulation guidance provided for in vitro and in vivo use.
Catalog No. 50-003-03208 Supplier Medchemexpress LLC Supplier No. HYW01524010MM/1ML
N-Acetyl-L-glutamic acid is an endogenous N-acyl derivative of L-glutamic acid used as a biochemical reagent and analytical standard. The product is available as a ready-to-use 10 mM solution in DMSO (1 mL) and is accompanied by molecular data and solubility guidance for laboratory use.
  • Endogenous metabolite involved in the urea cycle and metabolic assays.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and screening.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular weight 189.17 g/mol and formula C7H11NO5 for reference.
  • Recommended solvent and formulation guidance provided for in vitro and in vivo use.