Learn More
Cayman Chemical N-acetyl-L-Glutamine, Purity: ≥95%, CAS: 2490-97-3, 500 g
SDP
Catalog No. 502913365
Encompass_Preferred
An acetylated form of L-glutamine; decreases infarct volume and prevents neuronal apoptosis in the substantia nigra in a rat model of MCAO-induced cerebral ischemia-reperfusion injury; prevents decreases in body weight in protein-energy malnourished pigs at 8.28 g/animal per day
Catalog No. 50-291-3365 Supplier Cayman Chemical Supplier No. 35398500 G
An acetylated form of L-glutamine; decreases infarct volume and prevents neuronal apoptosis in the substantia nigra in a rat model of MCAO-induced cerebral ischemia-reperfusion injury; prevents decreases in body weight in protein-energy malnourished pigs at 8.28 g/animal per day