Learn More
Cayman Chemical N-acetyl-L-Isoleucine, Purity: ≥95%, CAS: 3077-46-1, 100 g
SDP
Catalog No. 502910153
Encompass_Preferred
An acetylated form of L-isoleucine; urine levels are increased in patients with MSUD
Catalog No. 50-291-0153 Supplier Cayman Chemical Supplier No. 35545100 G
An acetylated form of L-isoleucine; urine levels are increased in patients with MSUD