Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-L-leucyl-L-glutamyl-L-valyl-L-aspartyl aldehyde | 402832-01-3 | 98.4% | 500.54 g/mol | C22H36N4O9 | 5 MG
SDP

Catalog No. 5000221887
Encompass_Preferred

Ac-LEVD-CHO is an aldehyde peptide inhibitor of caspase-4 (sequence Ac-Leu-Glu-Val-Asp-CHO) used in biochemical and cell-based studies to inhibit caspase-4 activity. It is provided as a purified research reagent suitable for laboratory assays and mechanistic studies.

  • Peptide aldehyde inhibitor of caspase-4.
  • Sequence Ac-Leu-Glu-Val-Asp-CHO.
  • Purity 98.4%.
  • Molecular weight 500.54 g/mol.
  • Soluble in DMSO and water.
  • Available in small-mass vials for research use.

Catalog No. 50-002-21887 Supplier Medchemexpress LLC Supplier No. HY1367275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-LEVD-CHO is an aldehyde peptide inhibitor of caspase-4 (sequence Ac-Leu-Glu-Val-Asp-CHO) used in biochemical and cell-based studies to inhibit caspase-4 activity. It is provided as a purified research reagent suitable for laboratory assays and mechanistic studies.

  • Peptide aldehyde inhibitor of caspase-4.
  • Sequence Ac-Leu-Glu-Val-Asp-CHO.
  • Purity 98.4%.
  • Molecular weight 500.54 g/mol.
  • Soluble in DMSO and water.
  • Available in small-mass vials for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.