Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-ε-Acetyl-L-Lysine, MP Biomedicals™

Catalog No. MP021502352 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021502352 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021502352 Supplier MP Biomedicals, Inc Supplier No. 02150235.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used together with other lysine analogues. N-ε-Acetyl-L-Lysine, MP Biomedicals (L-AcK) is an R-chain N-acetylated α amino acid used to differentiate and characterize various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 692-04-6
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00002639
InChI Key DTERQYGMUDWYAZ-ZETCQYMHSA-N
Synonym n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine
PubChem CID 92832
ChEBI CHEBI:17752
IUPAC Name (2S)-2-amino-6-acetamidohexanoic acid
SMILES CC(=O)NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 247.1°C to 247.3°C (literature)
Quantity 250 mg
Solubility Information Soluble in 80% AcOH (5mg/mL, clear and colorless solution) and water.
Formula Weight 188.26
Percent Purity ≥99%
Grade Not Available
Physical Form Powder
Recommended Storage Store at –20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.