Learn More
Medchemexpress LLC HY-W012499 100mg Medchemexpress, N-Acetyl-L-methionine CAS:65-82-7 Purity:>98%
SDP
Catalog No. 501970689
Encompass_Preferred
Medchemexpress, HY-W012499 100mg N-Acetyl-L-methionine CAS:65-82-7 N-Acetyl-L-methionine, a human metabolite, is nutritionally and metabolically equivalent to L-methionine. L-methionine is an indispensable amino acid required for normal growth and development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0689 Supplier Medchemexpress LLC Supplier No. HYW012499100MG
Medchemexpress, HY-W012499 100mg N-Acetyl-L-methionine CAS:65-82-7 N-Acetyl-L-methionine, a human metabolite, is nutritionally and metabolically equivalent to L-methionine. L-methionine is an indispensable amino acid required for normal growth and development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.