Learn More
Medchemexpress LLC N-Acetyl-L-methionine | 65-82-7 | 99.9% | 191.25 | 1 ML
SDP
Catalog No. 5000452801
Encompass_Preferred
N-Acetyl-L-methionine is a dietary supplement that increases caspase-3 activity, inhibits mitochondrial activity, and promotes lactation while improving meat quality.
  • Reduces chemical modifications and structural changes of recombinant human serum albumin.
  • Inhibits the growth and mitochondrial activity of Jurkat and MTC-SK cells.
  • Preserves normal pharmacokinetics of indocyanine green-labeled rHSA in male mice.
  • Increases milk fat concentration and yield in lactating dairy cows.
  • Improves meat quality by enhancing antioxidant effects of lipids and proteins.
  • Affects weight gain in growing rats at higher dietary levels.
Catalog No. 50-004-52801 Supplier Medchemexpress LLC Supplier No. HYW01249910MM/1ML
N-Acetyl-L-methionine is a dietary supplement that increases caspase-3 activity, inhibits mitochondrial activity, and promotes lactation while improving meat quality.
  • Reduces chemical modifications and structural changes of recombinant human serum albumin.
  • Inhibits the growth and mitochondrial activity of Jurkat and MTC-SK cells.
  • Preserves normal pharmacokinetics of indocyanine green-labeled rHSA in male mice.
  • Increases milk fat concentration and yield in lactating dairy cows.
  • Improves meat quality by enhancing antioxidant effects of lipids and proteins.
  • Affects weight gain in growing rats at higher dietary levels.