Learn More
Selleck Chemical LLC N-Acetyl-L-methionine
SDP
Catalog No. 5000003171
Encompass_Preferred
N-Acetyl-L-methionine (Ac-Met-OH) is an L-methionine derivative that is nutritionally and metabolically equivalent to L-methionine It is a superior ROS scavenger
Catalog No. 50-000-03171 Supplier Selleck Chemical LLC Supplier No. S620525MG
N-Acetyl-L-methionine (Ac-Met-OH) is an L-methionine derivative that is nutritionally and metabolically equivalent to L-methionine It is a superior ROS scavenger