Learn More
Cayman Chemical N-Acetyl-L-phenylalanine, Purity: ≥98%, CAS: 2018-61-3, 25 g
SDP
Catalog No. 502911540
Encompass_Preferred
A metabolite of L-phenylalanine; formed from L-phenylalanine by gut microbiota; serum levels are increased in male COVID-19 patients; hippocampal levels are increased in postmortem brains isolated from patients with Alzheimer's disease; urinary levels are negatively correlated with phthalate exposure in normal weight children
Catalog No. 50-291-1540 Supplier Cayman Chemical Supplier No. 3625325 G
A metabolite of L-phenylalanine; formed from L-phenylalanine by gut microbiota; serum levels are increased in male COVID-19 patients; hippocampal levels are increased in postmortem brains isolated from patients with Alzheimer's disease; urinary levels are negatively correlated with phthalate exposure in normal weight children