Learn More
Cayman Chemical N-Acetyl-L-thyroxine, Purity: ≥98%, CAS: 26041-51-0, 500 mg
SDP
Catalog No. 502910929
Encompass_Preferred
N-Acetyl-L-thyroxine is an acetylated derivative of L-thyroxine (Item No. 14116).{61994} It only weakly binds to thyroxine-binding globulin (TBG) in isolated human serum with a relative binding affinity of 25 compared with L-thyroxine. N-Acetyl-L-thyroxine is also a potential impurity in commercial preparations of L-thyroxine.{61995}
Catalog No. 50-291-0929 Supplier Cayman Chemical Supplier No. 35197500 MG
N-Acetyl-L-thyroxine is an acetylated derivative of L-thyroxine (Item No. 14116).{61994} It only weakly binds to thyroxine-binding globulin (TBG) in isolated human serum with a relative binding affinity of 25 compared with L-thyroxine. N-Acetyl-L-thyroxine is also a potential impurity in commercial preparations of L-thyroxine.{61995}