Learn More
Medchemexpress LLC N-Acetyl-L-tryptophan | 1218-34-4 | 99.8% | 246.26 | 5 G
SDP
Catalog No. 5000452291
Encompass_Preferred
N-Acetyl-L-tryptophan is an antagonist of the neurokinin-1 receptor (NK-1R) that disrupts the binding of substance P (SP) to NK-1R, leading to neuroprotective effects and improved memory deficits and motor impairments. It also acts as an inhibitor of cytochrome c, exhibiting antioxidant and anti-inflammatory effects by inhibiting IL-1β expression and caspase-1 activation. This product is promising for research in neurodegenerative and inflammatory diseases.
  • Disrupts the binding of substance P (SP) to NK-1R.
  • Exhibits neuroprotective effects.
  • Improves memory deficits and motor impairments.
  • Acts as an inhibitor of cytochrome c.
  • Exhibits antioxidant and anti-inflammatory effects.
  • Inhibits IL-1β expression and caspase-1 activation.
  • Promising for research in neurodegenerative and inflammatory diseases.
Catalog No. 50-004-52291 Supplier Medchemexpress LLC Supplier No. HYW0119785G
N-Acetyl-L-tryptophan is an antagonist of the neurokinin-1 receptor (NK-1R) that disrupts the binding of substance P (SP) to NK-1R, leading to neuroprotective effects and improved memory deficits and motor impairments. It also acts as an inhibitor of cytochrome c, exhibiting antioxidant and anti-inflammatory effects by inhibiting IL-1β expression and caspase-1 activation. This product is promising for research in neurodegenerative and inflammatory diseases.
  • Disrupts the binding of substance P (SP) to NK-1R.
  • Exhibits neuroprotective effects.
  • Improves memory deficits and motor impairments.
  • Acts as an inhibitor of cytochrome c.
  • Exhibits antioxidant and anti-inflammatory effects.
  • Inhibits IL-1β expression and caspase-1 activation.
  • Promising for research in neurodegenerative and inflammatory diseases.