Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-tryptophan Ethyl Ester 98.0+%, TCI America™
SDP

Catalog No. A01221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A01221G Supplier TCI America Supplier No. A01221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2382-80-1
Molecular Formula C15H18N2O3
Molecular Weight (g/mol) 274.32
MDL Number MFCD00005643
InChI Key KQGQONPKSKUHHT-AWEZNQCLSA-N
Synonym Ac-Trp-OEt
PubChem CID 2724382
IUPAC Name ethyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
SMILES CCOC(=O)C(CC1=CNC2=CC=CC=C21)NC(=O)C

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 1 g
Formula Weight 274.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-tryptophan Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.