Learn More
Medchemexpress LLC N-acetyl-L-tryptophyl-L-leucyl-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-L-alaninamide | 1104011-59-7 | 99.8% | 587.67 | C32H37N5O6 | 5MG
SDP
Catalog No. 5000235734
Encompass_Preferred
Ac-WLA-AMC is a fluorogenic peptide substrate selective for the chymotrypsin-like (β5) subunit of the 20S constitutive proteasome. Upon enzymatic cleavage the 7-amino-4-methylcoumarin (AMC) fluorophore is released, enabling sensitive fluorometric measurement of proteasome β5 activity in biochemical assays. Supplied as a high-purity solid for research use.
Catalog No. 50-002-35734 Supplier Medchemexpress LLC Supplier No. HY1230515MG
Ac-WLA-AMC is a fluorogenic peptide substrate selective for the chymotrypsin-like (β5) subunit of the 20S constitutive proteasome. Upon enzymatic cleavage the 7-amino-4-methylcoumarin (AMC) fluorophore is released, enabling sensitive fluorometric measurement of proteasome β5 activity in biochemical assays. Supplied as a high-purity solid for research use.