Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-Tyrosine, MP Biomedicals™

Catalog No. ICN10015025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10015025 25 g
ICN10015005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10015025 Supplier MP Biomedicals, Inc Supplier No. 0210015025

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 537-55-3
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00037190
InChI Key CAHKINHBCWCHCF-UEQNJFAPNA-N
Synonym n-acetyl-l-tyrosine, ac-tyr-oh, acetyl-l-tyrosine, n-acetyl-tyrosine, n-acetyltyrosine, acetyl tyrosine, l-n-acetyltyrosine, tyrosine, n-acetyl, l-tyrosine, n-acetyl, unii-da8g610zo5
PubChem CID 68310
ChEBI CHEBI:21563
IUPAC Name (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Quantity 25 g
Formula Weight 223.2
Physical Form Crystalline
Chemical Name or Material N-Acetyl-L-Tyrosine
EINECSNumber 208-671-3
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.