Learn More
Medchemexpress LLC N-Acetyl-L-tyrosine (Standard) | 537-55-3 | 99.5% | 223.23 | 10 MG
SDP
Catalog No. 5000452808
Encompass_Preferred
N-Acetyl-L-tyrosine is an orally active endogenous mitochondrial stress response regulator that can permeate the cell membrane by passive diffusion. It induces low-level reactive oxygen species (ROS) generation by transiently perturbing mitochondrial membrane potential, triggering reverse signaling to activate FoxO and Keap1 pathways.
  • Enhances expression of antioxidant enzyme genes
  • Exerts anti-stress and cytoprotective effects
  • Can improve heat stress tolerance
  • Inhibits tumor growth
  • Regulates energy metabolism
  • Used in research of aging, metabolic diseases (such as diabetes), and cancer
Catalog No. 50-004-52808 Supplier Medchemexpress LLC Supplier No. HYW012382R10MG
N-Acetyl-L-tyrosine is an orally active endogenous mitochondrial stress response regulator that can permeate the cell membrane by passive diffusion. It induces low-level reactive oxygen species (ROS) generation by transiently perturbing mitochondrial membrane potential, triggering reverse signaling to activate FoxO and Keap1 pathways.
  • Enhances expression of antioxidant enzyme genes
  • Exerts anti-stress and cytoprotective effects
  • Can improve heat stress tolerance
  • Inhibits tumor growth
  • Regulates energy metabolism
  • Used in research of aging, metabolic diseases (such as diabetes), and cancer