Learn More
Medchemexpress LLC N-acetyl-L-tyrosyl-L-valyl-N-[(1S)-2-carboxyl-1-formylethyl]-L-alaninamide | 143313-51-3 | 98.2% | 5 MG
SDP
Catalog No. 5000234068
Encompass_Preferred
Ac-YVAD-CHO is an N-acetyl tetrapeptide aldehyde that selectively and reversibly inhibits caspase-1 (interleukin-1β converting enzyme). It is used in biochemical and cellular research to block caspase-1-mediated processes and is supplied as a high-purity powder suitable for assay development.
  • Potent and selective caspase-1 inhibitor.
  • Reversible tetrapeptide aldehyde mechanism of action.
  • High purity suitable for research applications.
  • Available in small-scale quantities for assay and biochemical studies.
  • Molecular weight 492.52 g·mol⁻¹; formula C23H32N4O8.
  • For research use only; follow applicable safety protocols.
Catalog No. 50-002-34068 Supplier Medchemexpress LLC Supplier No. HY1200195MG
Ac-YVAD-CHO is an N-acetyl tetrapeptide aldehyde that selectively and reversibly inhibits caspase-1 (interleukin-1β converting enzyme). It is used in biochemical and cellular research to block caspase-1-mediated processes and is supplied as a high-purity powder suitable for assay development.
  • Potent and selective caspase-1 inhibitor.
  • Reversible tetrapeptide aldehyde mechanism of action.
  • High purity suitable for research applications.
  • Available in small-scale quantities for assay and biochemical studies.
  • Molecular weight 492.52 g·mol⁻¹; formula C23H32N4O8.
  • For research use only; follow applicable safety protocols.