Learn More
Medchemexpress LLC N-acetyl-L-valine | 96-81-1 | MFCD00066066 | 98.0% | 159.19 g/mol | C7H13NO3 | 250 MG
SDP
Catalog No. 5000302782
Encompass_Preferred
N-acetyl-L-valine is an N-acetylated derivative of the amino acid valine supplied as an analytical standard for research and metabolite analysis. It is commonly used as an endogenous metabolite marker and can assist in detecting metabolic disorders such as maple syrup urine disease (MSUD).
  • High purity for analytical applications: 98.0%.
  • Molecular formula C7H13NO3 and molecular weight 159.19 g/mol.
  • Suitable for use as an analytical standard and metabolite reference.
  • Available in multiple sizes, including a 250 mg solid option.
  • Provided with a supporting datasheet for analytical characterization.
Catalog No. 50-003-02782 Supplier Medchemexpress LLC Supplier No. 250MGHYW015466
N-acetyl-L-valine is an N-acetylated derivative of the amino acid valine supplied as an analytical standard for research and metabolite analysis. It is commonly used as an endogenous metabolite marker and can assist in detecting metabolic disorders such as maple syrup urine disease (MSUD).
  • High purity for analytical applications: 98.0%.
  • Molecular formula C7H13NO3 and molecular weight 159.19 g/mol.
  • Suitable for use as an analytical standard and metabolite reference.
  • Available in multiple sizes, including a 250 mg solid option.
  • Provided with a supporting datasheet for analytical characterization.