Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-L-valyl-L-alpha-glutamyl-L-isoleucyl-L-aspart-1-al | 00-00-0 | MFCD01318861 | 99.8% | 614.57 g/mol | C24H37F3N4O11 | 10 MG
SDP

Catalog No. 5000242360
Encompass_Preferred

Ac-VEID-CHO TFA is a peptide-derived reversible caspase inhibitor for biochemical research, commonly used to probe caspase activity and mechanisms relevant to neurodegeneration. It is formulated as a salt suitable for in vitro enzymatic assays and VEIDase activity studies.

  • Peptide-derived reversible caspase inhibitor.
  • Biochemical IC50 values: caspase-3 13.6 nM, caspase-6 16.2 nM, caspase-7 162.1 nM.
  • High purity (≈99.8%).
  • White to off-white solid, soluble in water with sonication and warming.
  • Limited cellular accumulation; low activity in intact cell assays (>100 μM).
  • Recommended storage: sealed, -80°C for long term, -20°C for up to 1 year.

Catalog No. 50-002-42360 Supplier Medchemexpress LLC Supplier No. HY108312A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-VEID-CHO TFA is a peptide-derived reversible caspase inhibitor for biochemical research, commonly used to probe caspase activity and mechanisms relevant to neurodegeneration. It is formulated as a salt suitable for in vitro enzymatic assays and VEIDase activity studies.

  • Peptide-derived reversible caspase inhibitor.
  • Biochemical IC50 values: caspase-3 13.6 nM, caspase-6 16.2 nM, caspase-7 162.1 nM.
  • High purity (≈99.8%).
  • White to off-white solid, soluble in water with sonication and warming.
  • Limited cellular accumulation; low activity in intact cell assays (>100 μM).
  • Recommended storage: sealed, -80°C for long term, -20°C for up to 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.