Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-L-valyl-L-alpha-glutamyl-L-isoleucyl-L-aspart-1-al | 00-00-0 | MFCD01318861 | 99.8% | 614.57 g/mol | C24H37F3N4O11 | 5 MG
SDP

Catalog No. 5000243438
Encompass_Preferred

Ac-VEID-CHO TFA is a peptide-derived aldehyde inhibitor of caspases used in biochemical research to study apoptosis and neurodegenerative disease mechanisms. The trifluoroacetate salt is supplied as a white to off-white solid with high purity and is suitable for assays requiring nanomolar potency against executioner caspases.

  • Purity 99.8% by HPLC.
  • Potent inhibition of caspase-3, caspase-6, and caspase-7 (IC50: 13.6 nM, 16.2 nM, 162.1 nM).
  • Supplied as a solid, white to off-white powder.
  • Soluble in water at approximately 1 mg/mL with sonication and warming.
  • Storage recommendations include cold, sealed conditions to preserve stability.
  • Designed for research applications in apoptosis and neurodegenerative disease models.
  • Available in small research-scale quantities for assay development.

Catalog No. 50-002-43438 Supplier Medchemexpress LLC Supplier No. HY108312A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-VEID-CHO TFA is a peptide-derived aldehyde inhibitor of caspases used in biochemical research to study apoptosis and neurodegenerative disease mechanisms. The trifluoroacetate salt is supplied as a white to off-white solid with high purity and is suitable for assays requiring nanomolar potency against executioner caspases.

  • Purity 99.8% by HPLC.
  • Potent inhibition of caspase-3, caspase-6, and caspase-7 (IC50: 13.6 nM, 16.2 nM, 162.1 nM).
  • Supplied as a solid, white to off-white powder.
  • Soluble in water at approximately 1 mg/mL with sonication and warming.
  • Storage recommendations include cold, sealed conditions to preserve stability.
  • Designed for research applications in apoptosis and neurodegenerative disease models.
  • Available in small research-scale quantities for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.