Learn More
Medchemexpress LLC N-Acetyl [Leu28, Leu31] neuropeptide Y (24-36) | 155709-24-3 | 98.0% | 1787.08 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1014810MG
Encompass_Preferred
N-Acetyl [Leu28, Leu31] neuropeptide Y (24-36) is a selective agonist of the neuropeptide Y Y2 receptor. It has been observed to attenuate cardiac vagal action in anesthetized rats. This product is for research use only.
  • Selective agonist of neuropeptide Y Y2 receptor
  • Attenuates cardiac vagal action in anesthetized rats
  • For research use only
Catalog No. 50-004-23241