Learn More
Medchemexpress LLC N-Acetyl [Leu28, Leu31] neuropeptide Y (24-36) | 155709-24-3 | 98.0% | 1787.08 | 5 MG

Supplier: Medchemexpress LLC HYP101485MG
N-Acetyl [Leu28, Leu31] neuropeptide Y (24-36) is a selective agonist of the neuropeptide Y Y2 receptor. It attenuates cardiac vagal action in anesthetized rats and is for research use only.
- Selective agonist of neuropeptide Y Y2 receptor
- Attenuates cardiac vagal action in anesthetized rats
- Solid appearance, white to off-white color
- Molecular formula: C81H131N27O19
- Sequence: Ac-Leu-Arg-His-Tyr-Leu-Asn-Leu-Leu-Thr-Arg-Gln-Arg-Tyr-NH2
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.