Learn More
Cayman Chemical N-acetyl LeukotrIen E4 100ug
SDP
Catalog No. 502981550
Encompass_Preferred
N-acetyl LTE4 is the major inactive metabolite of LTE4 found in bile. This route of metabolism is prominent in the rat, but of minor importance in humans.{544,546} N-acetyl LTE4 is 100 times less potent than LTC4 as a vasoconstricting agent.{546} In healthy human subjects urinary excretion of N-acetyl LTE4 is about 1.5 nmol/mol creatinine, which is considerably less than that of LTE4 (12 nmol/mol creatinine).{307}
Catalog No. 50-298-1550 Supplier Cayman Chemical Supplier No. 20420100 UG
N-acetyl LTE4 is the major inactive metabolite of LTE4 found in bile. This route of metabolism is prominent in the rat, but of minor importance in humans.{544,546} N-acetyl LTE4 is 100 times less potent than LTC4 as a vasoconstricting agent.{546} In healthy human subjects urinary excretion of N-acetyl LTE4 is about 1.5 nmol/mol creatinine, which is considerably less than that of LTE4 (12 nmol/mol creatinine).{307}