Learn More
Medchemexpress LLC N-acetyl mesalazine | 51-59-2 | 99.9% | C9H9NO4 | 5 MG
SDP
Catalog No. 5000434707
Encompass_Preferred
N-Acetyl mesalazine (N-Acetyl-5-aminosalicylic acid) is the primary intestinal metabolite of 5-Aminosalicylic Acid and serves as a biomarker for evaluating its efficacy. It can scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation. This compound can be used in the study of diseases such as colitis and colon cancer.
  • Primary intestinal metabolite of 5-aminosalicylic acid
  • Serves as a biomarker for evaluating efficacy of 5-aminosalicylic acid
  • Scavenges free radicals
  • Reduces DNA base hydroxylation
  • Ameliorates mucosal inflammation
  • Can be used in the study of diseases such as colitis and colon cancer
  • Inhibits OH radical-stimulated DNA base hydroxylation in calf thymus DNA
Catalog No. 50-004-34707 Supplier Medchemexpress LLC Supplier No. HY66008S5MG
N-Acetyl mesalazine (N-Acetyl-5-aminosalicylic acid) is the primary intestinal metabolite of 5-Aminosalicylic Acid and serves as a biomarker for evaluating its efficacy. It can scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation. This compound can be used in the study of diseases such as colitis and colon cancer.
  • Primary intestinal metabolite of 5-aminosalicylic acid
  • Serves as a biomarker for evaluating efficacy of 5-aminosalicylic acid
  • Scavenges free radicals
  • Reduces DNA base hydroxylation
  • Ameliorates mucosal inflammation
  • Can be used in the study of diseases such as colitis and colon cancer
  • Inhibits OH radical-stimulated DNA base hydroxylation in calf thymus DNA