Learn More
Medchemexpress LLC N-Acetyl mesalazine | 51-59-2 | 99.9% | C9H9NO4 | 50 MG
SDP
Catalog No. 5000433825
Encompass_Preferred
N-Acetyl mesalazine is the primary intestinal metabolite of 5-Aminosalicylic Acid and serves as a biomarker for evaluating the efficacy of 5-Aminosalicylic Acid. It is utilized in the study of diseases such as colitis and colon cancer, demonstrating its ability to scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation.
  • Functions as a biomarker for 5-Aminosalicylic Acid efficacy
  • Scavenges free radicals and reduces DNA base hydroxylation
  • Ameliorates mucosal inflammation
  • Useful for studying diseases like colitis and colon cancer
Catalog No. 50-004-33825 Supplier Medchemexpress LLC Supplier No. HY6600850MG
N-Acetyl mesalazine is the primary intestinal metabolite of 5-Aminosalicylic Acid and serves as a biomarker for evaluating the efficacy of 5-Aminosalicylic Acid. It is utilized in the study of diseases such as colitis and colon cancer, demonstrating its ability to scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation.
  • Functions as a biomarker for 5-Aminosalicylic Acid efficacy
  • Scavenges free radicals and reduces DNA base hydroxylation
  • Ameliorates mucosal inflammation
  • Useful for studying diseases like colitis and colon cancer