Learn More
Medchemexpress LLC N-acetyl mesalazine | 51-59-2 | MFCD00463101 | 99.9% | 195.17 | C9H9NO4 | 10 MG
SDP
Catalog No. 5000298728
Encompass_Preferred
N-acetyl mesalazine (N-acetyl-5-aminosalicylic acid) is the primary intestinal metabolite of 5-aminosalicylic acid supplied as an analytical standard for laboratory research. It is used in metabolic, pharmacokinetic, and analytical method development studies, and is intended for research use only.
  • Analytical standard for research use only.
  • High purity: 99.9%.
  • Available as solid milligram quantities and 10 mM DMSO solutions.
  • Useful in metabolic and pharmacokinetic studies of 5-ASA.
  • Documented chemical identifiers and safety data available.
Catalog No. 50-002-98728 Supplier Medchemexpress LLC Supplier No. HY6600810MG
N-acetyl mesalazine (N-acetyl-5-aminosalicylic acid) is the primary intestinal metabolite of 5-aminosalicylic acid supplied as an analytical standard for laboratory research. It is used in metabolic, pharmacokinetic, and analytical method development studies, and is intended for research use only.
  • Analytical standard for research use only.
  • High purity: 99.9%.
  • Available as solid milligram quantities and 10 mM DMSO solutions.
  • Useful in metabolic and pharmacokinetic studies of 5-ASA.
  • Documented chemical identifiers and safety data available.