Learn More
CPC Scientific Ac-Nle-Glu-His-DPhe-Arg-Trp-Gly-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MSHS004A
Encompass
ONE-LETTER SEQUENCE: Ac-Nle-EH-DPhe-RWG-NH2MOLECULAR FORMULA: C47H64N14O10MOLECULAR WEIGHT:985.1STORAGE CONDITIONS: -20 5CSYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES: T.K. Sawyer et al., PNAS, 25, 1022 (1982)
Catalog No. 50-210-9839