Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-o-phenetidine 98.0+%, TCI America™
SDP

Catalog No. A010325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A010325G 25 g
A0103500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A010325G Supplier TCI America Supplier No. A010325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 581-08-8
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00059228
InChI Key SQRTWLGWCOJOTO-UHFFFAOYSA-N
PubChem CID 11383
IUPAC Name N-(2-ethoxyphenyl)acetamide
SMILES CCOC1=CC=CC=C1NC(=O)C

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Synonym N-Acetyl-2-ethoxyaniline, 2′C-Ethoxyacetanilide
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-o-phenetidine
EINECSNumber (3)-0695
RTECSNumber AM4350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.