Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-Pepstatin, Thermo Scientific™

Catalog No. AAJ66917MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66917MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66917MB Supplier Thermo Scientific Chemicals Supplier No. J66917MB

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28575-34-0
Molecular Formula C31H57N5O9
Molecular Weight (g/mol) 643.823
MDL Number MFCD00214071
InChI Key WKYBEGDEGRCZNF-LBTYKNIQSA-N
Synonym acetylpepstatin, acetyl-pepstatin, acetyl pepstatin, ac-val-val-sta-ala-sta, 3s,4s-4-2s-2-3s,4s-4-2s-2-2s-2-acetamido-3-methyl-butanoyl amino-3-methyl-butanoyl amino-3-hydroxy-6-methyl-heptanoyl amino propanoyl amino-3-hydroxy-6-methyl-heptanoic acid, pepstatin ac, pepsidin c, pepsidine c, 5hvp, n-acetyl pepstatin
PubChem CID 5481345
IUPAC Name (3S,4S)-4-[[(2S)-2-[[(3S,4S)-4-[[(2S)-2-[[(2S)-2-acetamido-3-methylbutanoyl]amino]-3-methylbutanoyl]amino]-3-hydroxy-6-methylheptanoyl]amino]propanoyl]amino]-3-hydroxy-6-methylheptanoic acid
SMILES CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)C)O

Specifications

Color White
Quantity 25 mg
Formula Weight 643.82
Physical Form Solid
Chemical Name or Material N-Acetyl-Pepstatin
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.