Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-phenylalanyl-lysyl-prolyl-leucyl-alanyl-alanyl-alanyl-arginine | 2891606-02-1 | 99.8% | C43H70N12O10 | 10MG
SDP

Catalog No. 5000259920
Encompass_Preferred

BM213 is a selective C5a receptor 1 (C5aR1) agonist peptide used in research to activate complement signaling, promote neutrophil extracellular trap formation, increase reactive oxygen species, and induce cardiomyocyte apoptosis; it is employed as a tool compound in myocardial ischemia/reperfusion injury studies.

  • Selective C5aR1 agonist with an EC50 of 59 nM.
  • Peptide sequence Ac-Phe-Lys-Pro-Leu-Ala-Ala-Ala-Arg (Ac-FKPLAAAR).
  • High reported purity (99.8%).
  • Molecular formula C43H70N12O10 and molecular weight 915.09.
  • Storage: protect from light and store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-59920 Supplier Medchemexpress LLC Supplier No. HY14523710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BM213 is a selective C5a receptor 1 (C5aR1) agonist peptide used in research to activate complement signaling, promote neutrophil extracellular trap formation, increase reactive oxygen species, and induce cardiomyocyte apoptosis; it is employed as a tool compound in myocardial ischemia/reperfusion injury studies.

  • Selective C5aR1 agonist with an EC50 of 59 nM.
  • Peptide sequence Ac-Phe-Lys-Pro-Leu-Ala-Ala-Ala-Arg (Ac-FKPLAAAR).
  • High reported purity (99.8%).
  • Molecular formula C43H70N12O10 and molecular weight 915.09.
  • Storage: protect from light and store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.