Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-S-benzyl-DL-cysteine 98.0+%, TCI America™
SDP

Catalog No. A07301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A07301G Supplier TCI America Supplier No. A07301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19538-71-7
Molecular Formula C12H15NO3S
Molecular Weight (g/mol) 253.32
MDL Number MFCD00067460
InChI Key BJUXDERNWYKSIQ-UHFFFAOYNA-N
Synonym n-acetyl-s-benzyl-dl-cysteine, n-acetyl-s-benzylcysteine, 2-acetamido-3-benzylthio propanoic acid, dl-benzylmercapturic acid, 3-benzylsulfanyl-2-acetamidopropanoic acid, cysteine,n-acetyl-s-phenylmethyl, acmc-209dmt, ac-dl-cys bzl-oh, enamine_000211, acmc-1bp0n
PubChem CID 561045
IUPAC Name 3-(benzylsulfanyl)-2-acetamidopropanoic acid
SMILES CC(=O)NC(CSCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 253.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-S-benzyl-DL-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.