Learn More
Medchemexpress LLC N-Acetyl-S-(N-methylcarbamoyl)-L-cysteine-d3 | 131685-10-4 | C7H9D3N2O4S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY144408S1MG
Encompass_Preferred
N-Acetyl-S-(N-methylcarbamoyl)-L-cysteine-d3 is the deuterium labeled N-Acetyl-S-(N-methylcarbamoyl)-L-cysteine. Stable heavy isotopes have been incorporated into drug molecules for use as tracers during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs. This product is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-54273