Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-trans-4-hydroxy-L-proline, 98%, Thermo Scientific Chemicals

Catalog No. AAB2376706
Change view
Click to view available options
Quantity:
2 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2376706 5 g
AAB2376704 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2376706 Supplier Thermo Scientific Chemicals Supplier No. B2376706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33996-33-7 | C7H11NO4 | 173.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33996-33-7
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
MDL Number MFCD00037339
InChI Key BAPRUDZDYCKSOQ-RITPCOANSA-N
Synonym oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum
PubChem CID 65784
IUPAC Name (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CC(CC1C(=O)O)O

Specifications

Melting Point 131°C to 133°C
Color White
Beilstein 84043
Merck Index 14,6903
Quantity 5 g
Formula Weight 173.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-trans-4-hydroxy-L-proline
Hazard Category H318
Hazard Statement H318-H335-H315; Causes serious eye damage. May cause respiratory irritation. Causes skin irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 251-780-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.