Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyladenosine | 16265-37-5 | MFCD30575188 | 99.9% | 309.28 g·mol⁻¹ | C12H15N5O5 | 1 ML
SDP

Catalog No. 5000281548
Encompass_Preferred

N-acetyladenosine is an adenosine analog supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research use. It exhibits vasodilatory activity and has reported effects on cancer progression pathways. The product is provided with high reported purity and recommended storage conditions for both powder and solution forms.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied in a 1 mL vial.
  • Reported purity 99.86%.
  • CAS number 16265-37-5 for unambiguous identification.
  • Chemical formula C12H15N5O5 and molecular weight 309.28 g·mol⁻¹.
  • Intended for biochemical and pharmacological research applications.
  • Storage: powder at -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-81548 Supplier Medchemexpress LLC Supplier No. HYW40206410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-acetyladenosine is an adenosine analog supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research use. It exhibits vasodilatory activity and has reported effects on cancer progression pathways. The product is provided with high reported purity and recommended storage conditions for both powder and solution forms.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied in a 1 mL vial.
  • Reported purity 99.86%.
  • CAS number 16265-37-5 for unambiguous identification.
  • Chemical formula C12H15N5O5 and molecular weight 309.28 g·mol⁻¹.
  • Intended for biochemical and pharmacological research applications.
  • Storage: powder at -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.