Learn More
Medchemexpress LLC HY-133026 100mg Medchemexpress, N-Acetylcarnosine CAS:56353-15-2 Purity:>98%
SDP
Catalog No. 502081325
Encompass_Preferred
Medchemexpress, HY-133026 100mg N-Acetylcarnosine CAS:56353-15-2 N-Acetylcarnosine, a natural histidine-containing dipeptide, is a source of pharmacological principal L-carnosine. N-Acetylcarnosine is a potent ophthalmic drug in human cataracts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1325 Supplier Medchemexpress LLC Supplier No. HY133026100MG
Medchemexpress, HY-133026 100mg N-Acetylcarnosine CAS:56353-15-2 N-Acetylcarnosine, a natural histidine-containing dipeptide, is a source of pharmacological principal L-carnosine. N-Acetylcarnosine is a potent ophthalmic drug in human cataracts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.