Learn More
Medchemexpress LLC N-Acetylcysteine amide | 38520-57-9 | MFCD01696115 | 99.88% | 162.21 | 1 MG
SDP
Catalog No. 501968779
Encompass_Preferred
N-Acetylcysteine amide is a cell membranes and blood-brain barrier permeant thiol antioxidant and neuroprotective agent that reduces ROS production. It is for research use only.
  • Reduces ROS levels and lipid peroxidation
  • Restores GSH/GSSG ratio and antioxidant enzyme activities
  • Protects human brain microvascular endothelial cells from methamphetamine-induced cell death
  • Improved CNS bioavailability
  • Improves cortical sparing and functional outcome following traumatic brain injury
  • Reduces oxidative stress and improves mitochondrial bioenergetics
  • Maintains mitochondrial glutathione content
Catalog No. 50-196-8779 Supplier Medchemexpress LLC Supplier No. HY1102561MG
N-Acetylcysteine amide is a cell membranes and blood-brain barrier permeant thiol antioxidant and neuroprotective agent that reduces ROS production. It is for research use only.
  • Reduces ROS levels and lipid peroxidation
  • Restores GSH/GSSG ratio and antioxidant enzyme activities
  • Protects human brain microvascular endothelial cells from methamphetamine-induced cell death
  • Improved CNS bioavailability
  • Improves cortical sparing and functional outcome following traumatic brain injury
  • Reduces oxidative stress and improves mitochondrial bioenergetics
  • Maintains mitochondrial glutathione content