Learn More
Medchemexpress LLC N-Acetyldesloratadin 1mg | 117796-52-8 | 1 MG
SDP
Catalog No. 5000227932
Encompass_Preferred
N-acetyldesloratadine is a research reagent that functions as a dual antagonist of platelet-activating factor (PAF) and histamine. It inhibits PAF-induced aggregation of human platelets (IC50 ≈ 0.6 μM) and is used in studies of allergic disease, platelet aggregation, and inflammation-related signaling. The compound is a white to off-white solid with molecular formula C21H21ClN2O and molecular weight 352.86.
Catalog No. 50-002-27932 Supplier Medchemexpress LLC Supplier No. HY1161201MG
N-acetyldesloratadine is a research reagent that functions as a dual antagonist of platelet-activating factor (PAF) and histamine. It inhibits PAF-induced aggregation of human platelets (IC50 ≈ 0.6 μM) and is used in studies of allergic disease, platelet aggregation, and inflammation-related signaling. The compound is a white to off-white solid with molecular formula C21H21ClN2O and molecular weight 352.86.