Learn More
Medchemexpress LLC N-acetyldopamine | 2494-12-4 | 250 MG
SDP
Catalog No. 5000421920
Encompass_Preferred
N-acetyldopamine is a sepiapterin reductase inhibitor and a catecholamine utilized by insects as sclerotizing precursors to harden their cuticle. It can attenuate LPS-stimulated TNF-α production and superoxide production in THP-1 cells. It is intended for research use only.
  • Sepiapterin reductase inhibitor
  • Catecholamine utilized by insects
  • Attenuates LPS-stimulated TNF-α production
  • Attenuates superoxide production in THP-1 cells
  • Targets endogenous metabolite, TNF receptor, reactive oxygen species (ROS), and reference standards
Catalog No. 50-004-21920 Supplier Medchemexpress LLC Supplier No. 250MGHYN7493
N-acetyldopamine is a sepiapterin reductase inhibitor and a catecholamine utilized by insects as sclerotizing precursors to harden their cuticle. It can attenuate LPS-stimulated TNF-α production and superoxide production in THP-1 cells. It is intended for research use only.
  • Sepiapterin reductase inhibitor
  • Catecholamine utilized by insects
  • Attenuates LPS-stimulated TNF-α production
  • Attenuates superoxide production in THP-1 cells
  • Targets endogenous metabolite, TNF receptor, reactive oxygen species (ROS), and reference standards