Learn More
Selleck Chemical LLC N-Acetylglucosamine-25mg
SDP
Supplier:  Selleck Chemical LLC S625725MG
Encompass_Preferred
N-Acetylglucosamine (GlcNAc) is a monosaccharide that usually polymerizes linearly through (1,4)-β-linkages. It may be helpful for the treatment of autoimmune diseases.
Catalog No. 50-239-0022