Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Acetylglycine

Catalog No. A16300100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16300100G 1 g
A163005G 500 g
A16300500G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A16300100G Supplier Sigma Aldrich Supplier No. A16300100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Acetylglycine

Specifications

CAS 34369-07-8
Melting Point 176°C
Molecular Formula C10H14N5Na2O13P3 -+ xH2O
Linear Formula C10H14N5Na2O13P3 -+ xH2O
MDL Number MFCD00150755
Quantity 1 g
Synonym Acetamidoacetic acid; Aceturic acid
Molecular Weight (g/mol) 551.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.