Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylglycine Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022750 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2750 Supplier MilliporeSigma Supelco Supplier No. 6956350MG

missing translation for 'validMainframeMsg'

N-Acetylglycine, belongs to the class of α-amino acids, which reportedly creates stable salts when combined with organic bases.

TRUSTED_SUSTAINABILITY

Specifications

CAS 543-24-8
Melting Point 207°C to 209°C (literature)
Flash Point Not applicable
Molecular Formula C4H7NO3
Linear Formula CH3CONHCH2CO2H
MDL Number MFCD00004275
Quantity 50 mg
Synonym Acetamidoacetic Acid; Aceturic Acid
Formula Weight 117.10
Percent Purity 98% to 102% (T)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.