Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylglycine 99.0+%, TCI America™
SDP

Catalog No. A0093100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0093100G 100 g
A009325G 25 g
A0093500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0093100G Supplier TCI America Supplier No. A0093100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 543-24-8
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD00004275
InChI Key OKJIRPAQVSHGFK-UHFFFAOYSA-N
Synonym n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid
PubChem CID 10972
ChEBI CHEBI:40410
IUPAC Name 2-acetamidoacetic acid
SMILES CC(=O)NCC(=O)O

Specifications

Melting Point 207°C
Color White
Quantity 100 g
Formula Weight 117.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylglycine
EINECSNumber (2)-1204
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.